6-(Ethylamino)-5-nitroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2424-5G HC-2424-1G
COBBHC-2424-5GEA 0 GBP
COBBHC-2424-5G COBBHC-2424-1G
6-(Ethylamino)-5-nitroquinoline
6-(Ethylamino)-5-nitroquinoline
Formula: C₁₁H₁₁N₃O₂
MDL Number: MFCD11855902
CAS Number: 99601-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR