6-(Trifluoromethoxy)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2355-5G HC-2355-1G
COBBHC-2355-5GEA 0 GBP
COBBHC-2355-5G COBBHC-2355-1G
6-(Trifluoromethoxy)quinoline
6-(Trifluoromethoxy)quinoline
Formula: C₁₀H₆F₃NO
MDL Number: MFCD11855894
CAS Number: 212695-45-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR