5-Amino-8-fluoroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 8-Fluoroquinolin-5-amine

HC-2225-1G HC-2225-250M
COBBHC-2225-1GEA 0 GBP
COBBHC-2225-1G COBBHC-2225-250M
5-Amino-8-fluoroquinoline
5-Amino-8-fluoroquinoline
Formula: C₉H₇FN₂
MW: 162.1637 g/mol
MDL Number: MFCD09042730
CAS Number: 175229-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR