6,8-Difluoroquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6969-5G
HC-6969-1G
COBBHC-6969-5GEA
0
GBP
COBBHC-6969-5G
COBBHC-6969-1G
6,8-Difluoroquinoline
6,8-Difluoroquinoline
Formula:
C₉H₅F₂N MW: 165.1400 g/mol |
MDL Number:
MFCD03662296 CAS Number: 145241-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...