2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008333-1G
008333-5G
FLUO008333-1GEA
0
GBP
FLUO008333-1G
FLUO008333-5G
2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile
2-[(4-Chlorobenzyl)oxy]-6-fluorobenzonitrile
Formula:
C₁₄H₉ClFNO |
MDL Number:
MFCD00068205 CAS Number: 175204-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...