3'-Formylbiphenyl-3-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4857-5G
CA-4857-1G
COBBCA-4857-5GEA
0
GBP
COBBCA-4857-5G
COBBCA-4857-1G
3'-Formylbiphenyl-3-carboxylic acid
3'-Formylbiphenyl-3-carboxylic acid
Formula:
C₁₄H₁₀O₃ |
MDL Number:
MFCD04039068 CAS Number: 222180-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...