3-Bromo-2-(3'-bromobenzyloxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8058-1G BB-8058-250M
COBBBB-8058-1GEA 0 GBP
COBBBB-8058-1G COBBBB-8058-250M
3-Bromo-2-(3'-bromobenzyloxy)phenylboronic acid
3-Bromo-2-(3'-bromobenzyloxy)phenylboronic acid
Formula: C₁₃H₁₁BBr₂O₃
MDL Number: MFCD06411319
CAS Number: 849062-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR