4'-Bromobiphenyl-4-ylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4'-Bromo-4-biphenylboronic acid

BB-8355-5G BB-8355-1G
COBBBB-8355-5GEA 0 GBP
COBBBB-8355-5G COBBBB-8355-1G
4'-Bromobiphenyl-4-ylboronic acid
4'-Bromobiphenyl-4-ylboronic acid
Formula: C₁₂H₁₀BBrO₂
MW: 276.9216 g/mol
MDL Number: MFCD02251652
CAS Number: 480996-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR