(1-(4-Bromophenyl)cyclopropyl)methanol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1109-1G OT-1109-5G OT-1109-250M
COBBOT-1109-1GEA 0 GBP
COBBOT-1109-1G COBBOT-1109-5G COBBOT-1109-250M
(1-(4-Bromophenyl)cyclopropyl)methanol
(1-(4-Bromophenyl)cyclopropyl)methanol
Formula: C₁₀H₁₁BrO
MW: 227.1 g/mol
MDL Number: MFCD07374454
CAS Number: 98480-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR