4-Chloro-3,5-dimethylphenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethyl-4-chlorophenylboronic acid pinacol ester, 2-(4-Chloro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5541-5G PN-5541-1G
COBBPN-5541-5GEA 0 GBP
COBBPN-5541-5G COBBPN-5541-1G
4-Chloro-3,5-dimethylphenylboronic acid pinacol ester
4-Chloro-3,5-dimethylphenylboronic acid pinacol ester
Formula: C₁₄H₂₀BClO₂
MW: 266.571 g/mol
MDL Number: MFCD12405360
CAS Number: 1111096-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR