3-Bromo-2-(3'-bromobenzyloxy)-5-methylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-8219-1G
BB-8219-250M
COBBBB-8219-1GEA
0
GBP
COBBBB-8219-1G
COBBBB-8219-250M
3-Bromo-2-(3'-bromobenzyloxy)-5-methylphenylboronic acid
3-Bromo-2-(3'-bromobenzyloxy)-5-methylphenylboronic acid
Formula:
C₁₄H₁₃BBr₂O₃ |
MDL Number:
MFCD06411363 CAS Number: 849052-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...