N,4-Dimethylaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2862-5G HC-2862-25G
COBBHC-2862-5GEA 0 GBP
COBBHC-2862-5G COBBHC-2862-25G
N,4-Dimethylaniline hydrochloride
N,4-Dimethylaniline hydrochloride
Formula: C₈H₁₂ClN
MDL Number: MFCD12913958
CAS Number: 2739-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR