N,2-Dimethylaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2861-25G
HC-2861-5G
COBBHC-2861-25GEA
0
GBP
COBBHC-2861-25G
COBBHC-2861-5G
N,2-Dimethylaniline hydrochloride
N,2-Dimethylaniline hydrochloride
Formula:
C₈H₁₂ClN |
MDL Number:
MFCD12913957 CAS Number: 10541-29-4 EINECS: 234-119-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...