5-Methoxy-2-methyl-4-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1456-5G
AN-1456-1G
COBBAN-1456-5GEA
0
GBP
COBBAN-1456-5G
COBBAN-1456-1G
5-Methoxy-2-methyl-4-nitroaniline
5-Methoxy-2-methyl-4-nitroaniline
Formula:
C₈H₁₀N₂O₃ |
MDL Number:
MFCD00010136 CAS Number: 106579-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...