N-Boc-4-iodo-3-methylaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-2562-5G AN-2562-1G
COBBAN-2562-5GEA 0 GBP
COBBAN-2562-5G COBBAN-2562-1G
N-Boc-4-iodo-3-methylaniline
N-Boc-4-iodo-3-methylaniline
Formula: C₁₂H₁₆INO₂
MDL Number: MFCD16036101
CAS Number: 1221793-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR