Methyl-4-(hydroxymethyl)benzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-(hydroxymethyl)benzoate

AS25237G100 AS25237G25
ACTIAS25237G100EA 0 GBP
ACTIAS25237G100 ACTIAS25237G25
Methyl-4-(hydroxymethyl)benzoate
Methyl 4-(hydroxymethyl)benzoate
Formula: C₉H₁₀O₃
MDL Number: MFCD00009984
CAS Number: 6908-41-4
EINECS: 230-017-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR