Methyl-4-methylsalicylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21406G25 AS21406G5
ACTIAS21406G25EA 0 GBP
ACTIAS21406G25 ACTIAS21406G5
Methyl-4-methylsalicylate
Methyl 4-methylsalicylate
Formula: C₉H₁₀O₃
MDL Number: MFCD00020130
CAS Number: 4670-56-8
EINECS: 225-117-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR