N-Boc-D-phenylalaninol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Boc-D-Phenylalaninol

AS20991G100 AS20991G25 AS20991G5
ACTIAS20991G100EA 0 GBP
ACTIAS20991G100 ACTIAS20991G25 ACTIAS20991G5
N-Boc-D-phenylalaninol
N-Boc-D-phenylalaninol
Formula: C₁₄H₂₁NO₃
MDL Number: MFCD00216472
CAS Number: 106454-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR