2-Amino-4'-(dimethylamino)acetophenone hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-1-[4-(dimethylamino)phenyl]ethan-1-one hydrochloride

AM-1600-5G AM-1600-1G
COBBAM-1600-5GEA 633.11 GBP
COBBAM-1600-5G COBBAM-1600-1G
2-Amino-4'-(dimethylamino)acetophenone hydrochloride
2-Amino-4'-(dimethylamino)acetophenone hydrochloride
Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD08689507
CAS Number: 152278-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR