3-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8182-1G BB-8182-250M
COBBBB-8182-1GEA 0 GBP
COBBBB-8182-1G COBBBB-8182-250M
3-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenylboronic acid
3-((2-Chloro-5-(trifluoromethyl)phenoxy)methyl)phenylboronic acid
Formula: C₁₄H₁₁BClF₃O₃
MDL Number: MFCD07784395
CAS Number: 871126-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR