4-((4-Chloro-3-methylphenoxy)methyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-((4-Chloro-3-methylphenoxy)methyl)phenyl)boronic acid

BB-8032-1G BB-8032-250M
COBBBB-8032-1GEA 0 GBP
COBBBB-8032-1G COBBBB-8032-250M
4-((4-Chloro-3-methylphenoxy)methyl)phenylboronic acid
4-((4-Chloro-3-methylphenoxy)methyl)phenylboronic acid
Formula: C₁₄H₁₄BClO₃
MW: 276.523 g/mol
MDL Number: MFCD14585463
CAS Number: 849052-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR