4-(Chloromethyl)phenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-2333-1G
PN-2333-5G
COBBPN-2333-1GEA
0
GBP
COBBPN-2333-1G
COBBPN-2333-5G
4-(Chloromethyl)phenylboronic acid pinacol ester
4-(Chloromethyl)phenylboronic acid pinacol ester
Formula:
C₁₃H₁₈BClO₂ |
MDL Number:
MFCD11053893 CAS Number: 1072945-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...