3-(Chloromethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(3-(Chloromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2478-1G PN-2478-5G
COBBPN-2478-1GEA 0 GBP
COBBPN-2478-1G COBBPN-2478-5G
3-(Chloromethyl)phenylboronic acid pinacol ester
3-(Chloromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₈BClO₂
MDL Number: MFCD11053847
CAS Number: 1073353-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR