5-Bromo-N,2-dimethylaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2860-5G HC-2860-1G
COBBHC-2860-5GEA 0 GBP
COBBHC-2860-5G COBBHC-2860-1G
5-Bromo-N,2-dimethylaniline hydrochloride
5-Bromo-N,2-dimethylaniline hydrochloride
Formula: C₈H₁₁BrClN
MDL Number: MFCD12756442
CAS Number: 1187386-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR