N-Boc-4-bromo-2-methylaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl (4-bromo-2-methylphenyl)carbamate, tert-Butyl 4-bromo-2-methylphenylcarbamate, 4-Bromo-2-methylaniline, N-BOC protected

AN-1569-5G AN-1569-1G
COBBAN-1569-5GEA 0 GBP
COBBAN-1569-5G COBBAN-1569-1G
N-Boc-4-bromo-2-methylaniline
N-Boc-4-bromo-2-methylaniline
Formula: C₁₂H₁₆BrNO₂
MW: 286.1700 g/mol
MDL Number: MFCD01765429
CAS Number: 306937-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR