N-Boc-5-bromo-2-methylaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: t-Butyl 5-bromo-2-methylphenylcarbamate

AN-1366-5G AN-1366-25G AN-1366-1G
COBBAN-1366-5GEA 0 GBP
COBBAN-1366-5G COBBAN-1366-25G COBBAN-1366-1G
N-Boc-5-bromo-2-methylaniline
N-Boc-5-bromo-2-methylaniline
Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD11504830
CAS Number: 221538-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR