2-(Aminocarbonylmethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-6042-1G PN-6042-250M
COBBPN-6042-1GEA 0 GBP
COBBPN-6042-1G COBBPN-6042-250M
2-(Aminocarbonylmethyl)phenylboronic acid pinacol ester
2-(Aminocarbonylmethyl)phenylboronic acid pinacol ester
Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD12913986
CAS Number: 1218789-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR