4-Nitro-N,N-diphenylaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Nitrotriphenylamine

AN-1322-1G AN-1322-5G
COBBAN-1322-1GEA 0 GBP
COBBAN-1322-1G COBBAN-1322-5G
4-Nitro-N,N-diphenylaniline
4-Nitro-N,N-diphenylaniline
Formula: C₁₈H₁₄N₂O₂
MW: 290.320 g/mol
MDL Number: MFCD00814232
CAS Number: 4316-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR