4-(Methylsulphonyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

32697-5G 32697-25G
FLUO32697-5GEA 0 GBP
FLUO32697-5G FLUO32697-25G
4-(Methylsulphonyl)aniline
4-(Methylsulphonyl)aniline
Formula: C₇H₉NO₂S
MDL Number: MFCD00025353
CAS Number: 5470-49-5
EINECS: 226-803-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR