N,N-Diethyl-2-(4-nitrophenoxy)ethanamine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1212-5G AN-1212-25G
COBBAN-1212-5GEA 0 GBP
COBBAN-1212-5G COBBAN-1212-25G
N,N-Diethyl-2-(4-nitrophenoxy)ethanamine
N,N-Diethyl-2-(4-nitrophenoxy)ethanamine
Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD01308350
CAS Number: 19881-36-8
EINECS: 243-394-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR