4-Thiocyanato-2-(trifluoromethoxy)aniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1399-5G AN-1399-1G
COBBAN-1399-5GEA 0 GBP
COBBAN-1399-5G COBBAN-1399-1G
4-Thiocyanato-2-(trifluoromethoxy)aniline
4-Thiocyanato-2-(trifluoromethoxy)aniline
Formula: C₈H₅F₃N₂OS
MDL Number: MFCD11855816
CAS Number: 1133115-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR