1-Iodo-2-nitro-4-(trifluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Iodo-5-(trifluoromethoxy)nitrobenzene

HC-2985-5G HC-2985-25G HC-2985-1G
COBBHC-2985-5GEA 0 GBP
COBBHC-2985-5G COBBHC-2985-25G COBBHC-2985-1G
1-Iodo-2-nitro-4-(trifluoromethoxy)benzene
1-Iodo-2-nitro-4-(trifluoromethoxy)benzene
Formula: C₇H₃F₃INO₃
MDL Number: MFCD04039213
CAS Number: 886762-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR