4-Amino-3-(trifluoromethoxy)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004025-1G 004025-5G
FLUO004025-1GEA 0 GBP
FLUO004025-1G FLUO004025-5G
4-Amino-3-(trifluoromethoxy)benzonitrile
4-Amino-3-(trifluoromethoxy)benzonitrile
Formula: C₈H₅F₃N₂O
MDL Number: MFCD00204177
CAS Number: 175278-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR