2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid,pinacol ester, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester

PN-2888-1G PN-2888-5G
COBBPN-2888-1GEA 0 GBP
COBBPN-2888-1G COBBPN-2888-5G
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₈BFO₄
MDL Number: MFCD11855962
CAS Number: 603122-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR