Methyl-4-(chlorosulphonyl)benzoate

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-chlorosulphonylbenzoate

ACTIAS21154G1EA 40 GBP
ACTIAS21154G1 ACTIAS21154G5
Methyl-4-(chlorosulphonyl)benzoate
Methyl 4-(chlorosulphonyl)benzoate
Formula: C₈H₇ClO₄S
MW: 234.6568 g/mol
MDL Number: MFCD00627554
CAS Number: 69812-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR