Methyl-3-(chlorosulphonyl)benzoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 3-(chlorosulphonyl)benzoate
AS20586G1
AS20586G25
AS20586G5
ACTIAS20586G1EA
16
GBP
ACTIAS20586G1
ACTIAS20586G25
ACTIAS20586G5
Methyl-3-(chlorosulphonyl)benzoate
Methyl 3-(chlorosulphonyl)benzoate
Formula:
C₈H₇ClO₄S MW: 234.6600 g/mol |
MDL Number:
MFCD06408800 CAS Number: 63555-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...