3-(4-Carbamoylphenoxy)propionic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22934G1
AS22934G5
ACTIAS22934G1EA
365
GBP
ACTIAS22934G1
ACTIAS22934G5
3-(4-Carbamoylphenoxy)propionic acid
3-(4-Carbamoylphenoxy)propionic acid
Formula:
C₁₀H₁₁NO₄ |
MDL Number:
MFCD09948835 CAS Number: 111140-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...