3-(2-Carbamoylphenoxy)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22935G1 AS22935G5
ACTIAS22935G1EA 0 GBP
ACTIAS22935G1 ACTIAS22935G5
3-(2-Carbamoylphenoxy)propionic acid
3-(2-Carbamoylphenoxy)propionic acid
Formula: C₁₀H₁₁NO₄
MDL Number: MFCD02364319
CAS Number: 103204-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR