3-Chloro-4-[(trifluoromethyl)thio]aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007810-5G 007810-1G
FLUO007810-5GEA 0 GBP
FLUO007810-5G FLUO007810-1G
3-Chloro-4-[(trifluoromethyl)thio]aniline
3-Chloro-4-[(trifluoromethyl)thio]aniline
Formula: C₇H₅ClF₃NS
MW: 227.64 g/mol
MDL Number: MFCD00190131
CAS Number: 64628-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR