2-(4-Chlorophenylthio)-6-fluorobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008334-1G
008334-5G
FLUO008334-1GEA
0
GBP
FLUO008334-1G
FLUO008334-5G
2-(4-Chlorophenylthio)-6-fluorobenzonitrile
2-(4-Chlorophenylthio)-6-fluorobenzonitrile
Formula:
C₁₃H₇ClFNS |
MDL Number:
MFCD00068208 CAS Number: 175204-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...