Phenyl-4-bromobenzenesulphonate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0514-5G
OT-0514-1G
OT-0514-25G
COBBOT-0514-5GEA
0
GBP
COBBOT-0514-5G
COBBOT-0514-1G
COBBOT-0514-25G
Phenyl-4-bromobenzenesulphonate
Phenyl 4-bromobenzenesulphonate
Formula:
C₁₂H₉BrO₃S |
MDL Number:
MFCD09475912 CAS Number: 5455-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...