4-Bromo-N,N-diphenylaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(4-Bromophenyl)diphenylamine, 4-Bromotriphenylamine
AN-1523-25G
AN-1523-1G
AN-1523-100G
AN-1523-5G
COBBAN-1523-25GEA
0
GBP
COBBAN-1523-25G
COBBAN-1523-1G
COBBAN-1523-100G
COBBAN-1523-5G
4-Bromo-N,N-diphenylaniline
4-Bromo-N,N-diphenylaniline
Formula:
C₁₈H₁₄BrN MW: 324.21 g/mol |
MDL Number:
MFCD01851266 CAS Number: 36809-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...