4,5-Dibromo-2-methoxy-N-methylaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1530-5G
AN-1530-1G
COBBAN-1530-5GEA
0
GBP
COBBAN-1530-5G
COBBAN-1530-1G
4,5-Dibromo-2-methoxy-N-methylaniline
4,5-Dibromo-2-methoxy-N-methylaniline
Formula:
C₈H₉Br₂NO |
MDL Number:
MFCD12913931 CAS Number: 1187386-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...