4-Bromo-N,N-dimethyl-2-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2736-5G HC-2736-1G
COBBHC-2736-5GEA 0 GBP
COBBHC-2736-5G COBBHC-2736-1G
4-Bromo-N,N-dimethyl-2-nitroaniline
4-Bromo-N,N-dimethyl-2-nitroaniline
Formula: C₈H₉BrN₂O₂
MW: 245.0700 g/mol
MDL Number: MFCD12546467
CAS Number: 829-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR