1-Bromo-2-fluoro-4-(methylsulphonyl)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0702-5G OT-0702-1G
COBBOT-0702-5GEA 0 GBP
COBBOT-0702-5G COBBOT-0702-1G
1-Bromo-2-fluoro-4-(methylsulphonyl)benzene
1-Bromo-2-fluoro-4-(methylsulphonyl)benzene
Formula: C₇H₆BrFO₂S
MDL Number: MFCD11053812
CAS Number: 1032825-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR