N-Formyl 3-bromo-2-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-2547-5G
AN-2547-1G
COBBAN-2547-5GEA
0
GBP
COBBAN-2547-5G
COBBAN-2547-1G
N-Formyl 3-bromo-2-nitroaniline
N-Formyl 3-bromo-2-nitroaniline
Formula:
C₇H₅BrN₂O₃ |
MDL Number:
MFCD13195639 CAS Number: 1215205-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...