2-(Morpholinosulphonyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-3835-5G
BB-3835-1G
COBBBB-3835-5GEA
0
GBP
COBBBB-3835-5G
COBBBB-3835-1G
2-(Morpholinosulphonyl)phenylboronic acid
2-(Morpholinosulphonyl)phenylboronic acid
Formula:
C₁₀H₁₄BNO₅S |
MDL Number:
MFCD09878327 CAS Number: 957062-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...