2-(Piperidin-1-ylsulphonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-(Piperidin-1-ylsulphonyl)phenyl)boronic acid

BB-3876-5G BB-3876-1G
COBBBB-3876-5GEA 0 GBP
COBBBB-3876-5G COBBBB-3876-1G
2-(Piperidin-1-ylsulphonyl)phenylboronic acid
2-(Piperidin-1-ylsulphonyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₄S
MW: 269.1300 g/mol
MDL Number: MFCD09475844
CAS Number: 957034-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR