4-(Tetrahydropyran-2-yloxy)phenylboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)phenylboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, (4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid
011039-1G
011039-5G
FLUO011039-1GEA
0
GBP
FLUO011039-1G
FLUO011039-5G
4-(Tetrahydropyran-2-yloxy)phenylboronic acid
4-(Tetrahydropyran-2-yloxy)phenylboronic acid
Formula:
C₁₁H₁₅BO₄ MW: 222.05 g/mol |
MDL Number:
MFCD02183525 CAS Number: 182281-01-2 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...