3,4-Bis(methoxycarbonyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(3,4-Bis(methoxycarbonyl)phenyl)boronic acid
BB-5363-1G
BB-5363-250M
COBBBB-5363-1GEA
0
GBP
COBBBB-5363-1G
COBBBB-5363-250M
3,4-Bis(methoxycarbonyl)phenylboronic acid
3,4-Bis(methoxycarbonyl)phenylboronic acid
Formula:
C₁₀H₁₁BO₆ MW: 238.0017 g/mol |
MDL Number:
MFCD11504854 CAS Number: 1072951-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...